Assay Details
Gene Symbol: nan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140439.3 | NP_648696.2 | 8 - 9 | 2556 | 81 | |
GenBank mRNA | AY262004.1 | - | 8 - 9 | 2557 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
