Assay Details
| Gene Symbol: bmm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169974.2 | NP_001163445.1 | 2 - 3 | 1176 | 73 | |
| NM_140466.2 | NP_648723.1 | 2 - 3 | 1176 | 73 | ||
| GenBank mRNA | AY051668.1 | - | 2 - 3 | 1121 | 73 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 bmm
bmm Genomic Map
Genomic Map Back To Top
 Back To Top