Assay Details
Gene Symbol: DCX-EMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274921.1 | NP_001261850.1 | 10 - 11 | 3510 | 53 | |
NM_001274922.1 | NP_001261851.1 | 10 - 11 | 2573 | 53 | ||
NM_140471.3 | NP_648728.2 | 10 - 11 | 3510 | 53 | ||
NM_140472.2 | NP_648729.1 | 10 - 11 | 2534 | 53 | ||
GenBank mRNA | BT030411.1 | - | 10 - 11 | 3106 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
