Assay Details
Gene Symbol: DCX-EMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274921.1 | NP_001261850.1 | 7 - 8 | 2295 | 110 | |
NM_001274922.1 | NP_001261851.1 | 7 - 8 | 1358 | 110 | ||
NM_140471.3 | NP_648728.2 | 7 - 8 | 2295 | 110 | ||
NM_140472.2 | NP_648729.1 | 7 - 8 | 1319 | 110 | ||
GenBank mRNA | BT030411.1 | - | 7 - 8 | 1891 | 110 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
