Assay Details
Gene Symbol: DCX-EMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274921.1 | NP_001261850.1 | 8 - 9 | 2699 | 76 | |
NM_001274922.1 | NP_001261851.1 | 8 - 9 | 1762 | 76 | ||
NM_140471.3 | NP_648728.2 | 8 - 9 | 2699 | 76 | ||
NM_140472.2 | NP_648729.1 | 8 - 9 | 1723 | 76 | ||
GenBank mRNA | BT030411.1 | - | 8 - 9 | 2295 | 76 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
