Assay Details
| Gene Symbol: Cpr72Ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140625.3 | NP_648882.1 | 1 | 1007 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr72Ea| Gene Symbol: Cpr72Ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140625.3 | NP_648882.1 | 1 | 1007 | 77 | |
Set Membership: |
|