Assay Details
Gene Symbol: RAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275014.1 | NP_001261943.1 | 2 - 3 | 570 | 84 | |
NM_140632.3 | NP_648889.2 | 2 - 3 | 500 | 84 | ||
NM_168664.2 | NP_730163.1 | 2 - 3 | 576 | 84 | ||
GenBank mRNA | BT021383.1 | - | 2 - 3 | 558 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
