Assay Details
Gene Symbol: Baldspot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140652.4 | NP_648909.1 | 4 - 5 | 640 | 100 | |
NM_168678.3 | NP_730187.2 | 4 - 5 | 700 | 100 | ||
GenBank mRNA | AF265296.1 | - | 4 - 5 | 652 | 100 | |
AY061121.1 | - | 4 - 5 | 652 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
