Assay Details
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300174.1 | NP_001287103.1 | 2 - 3 | 858 | 81 | |
NM_140759.3 | NP_649016.1 | 2 - 3 | 858 | 81 | ||
GenBank mRNA | AY069647.1 | - | 2 - 3 | 839 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
