Assay Details
Gene Symbol: Prestin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140767.2 | NP_649024.1 | 4 - 5 | 2418 | 76 | |
GenBank mRNA | BT133214.1 | - | 4 - 5 | 2386 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
