Assay Details
Gene Symbol: Prestin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140767.2 | NP_649024.1 | 5 - 6 | 2568 | 67 | |
GenBank mRNA | BT133214.1 | - | 5 - 6 | 2536 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
