Assay Details
| Gene Symbol: AstC-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031964.2 | NP_001027135.2 | 2 - 3 | 1577 | 72 | |
| NM_001275086.1 | NP_001262015.1 | 2 - 3 | 1577 | 72 | ||
| NM_001316469.1 | NP_001303398.1 | 2 - 3 | 1577 | 72 | ||
| NM_140782.4 | NP_649039.4 | 2 - 3 | 1577 | 72 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  G-protein coupled receptor  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 AstC-R2
AstC-R2 Genomic Map
Genomic Map Back To Top
 Back To Top