Assay Details
| Gene Symbol: AstC-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031964.2 | NP_001027135.2 | 3 - 4 | 1626 | 67 | |
| NM_001275086.1 | NP_001262015.1 | 3 - 4 | 1626 | 67 | ||
| NM_001316469.1 | NP_001303398.1 | 3 - 4 | 1626 | 67 | ||
| NM_140782.4 | NP_649039.4 | 3 - 4 | 1626 | 67 | ||
More Information
Set Membership: |
|

AstC-R2
Genomic Map