Assay Details
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140800.3 | NP_649057.1 | 1 - 2 | 120 | 103 | |
GenBank mRNA | AY102649.1 | - | 1 - 2 | 74 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
