Assay Details
| Gene Symbol: Cpr76Bd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104175.3 | NP_001097645.1 | 2 - 3 | 3450 | 73 | |
| NM_001275124.1 | NP_001262053.1 | 2 - 3 | 3450 | 73 | ||
More Information
Set Membership: |
|

Cpr76Bd
Genomic Map