Assay Details
Gene Symbol: TORIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275136.1 | NP_001262065.1 | 2 - 3 | 730 | 101 | |
NM_140892.4 | NP_649149.1 | 2 - 3 | 730 | 101 | ||
GenBank mRNA | AY061389.1 | - | 2 - 3 | 719 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
