Assay Details
| Gene Symbol: Tom20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275156.1 | NP_001262085.1 | 2 - 3 | 338 | 63 | |
| NM_140912.4 | NP_649169.3 | 1 - 2 | 212 | 63 | ||
| GenBank mRNA | AY061414.1 | - | 1 - 2 | 181 | 63 | |
More Information
Set Membership: |
|

Tom20
Genomic Map