Assay Details
Gene Symbol: CtIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300200.1 | NP_001287129.1 | 3 - 4 | 1432 | 83 | |
NM_140967.3 | NP_649224.1 | 3 - 4 | 1425 | 83 | ||
GenBank mRNA | AY071347.1 | - | 3 - 4 | 1418 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
