Assay Details
Gene Symbol: Vha68-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259086.2 | NP_001246015.1 | 4 - 5 | 1816 | 62 | |
NM_001273495.1 | NP_001260424.1 | 4 - 5 | 1846 | 62 | ||
NM_001273496.1 | NP_001260425.1 | 4 - 5 | 1900 | 62 | ||
NM_143747.3 | NP_652004.2 | 4 - 5 | 1858 | 62 | ||
NM_165021.2 | NP_723775.1 | 4 - 5 | 1836 | 62 | ||
NM_165022.2 | NP_723776.1 | 4 - 5 | 1903 | 62 | ||
GenBank mRNA | BT124897.1 | - | 4 - 5 | 1846 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
