Assay Details
| Gene Symbol: kuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057839.5 | NP_477187.1 | 11 - 12 | 4193 | 145 | |
| NM_165047.3 | NP_723825.1 | 11 - 12 | 4066 | 145 | ||
More Information
Set Membership: |
|

kuz
Genomic Map