Assay Details
Gene Symbol: NimB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165075.3 | NP_723857.1 | 1 - 2 | 515 | 66 | |
GenBank mRNA | BT012496.1 | - | 1 - 2 | 504 | 66 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
