Assay Details
| Gene Symbol: wb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201877.2 | NP_001188806.1 | 8 - 9 | 6522 | 85 | |
| NM_001201878.2 | NP_001188807.1 | 8 - 9 | 6604 | 85 | ||
| NM_001201879.2 | NP_001188808.1 | 8 - 9 | 6730 | 85 | ||
| NM_001201880.2 | NP_001188809.1 | 8 - 9 | 6543 | 85 | ||
| NM_057442.5 | NP_476790.3 | 10 - 11 | 8880 | 85 | ||
| NM_165086.3 | NP_723870.1 | 10 - 11 | 8880 | 85 | ||
More Information
Set Membership: |
|

wb
Genomic Map