Assay Details
Gene Symbol: ck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299020.1 | NP_001285949.1 | 11 - 12 | 6584 | 62 | |
NM_001299021.1 | NP_001285950.1 | 11 - 12 | 6747 | 62 | ||
NM_078847.4 | NP_523571.1 | 11 - 12 | 6579 | 62 | ||
NM_165099.3 | NP_723895.1 | 11 - 12 | 6584 | 62 | ||
GenBank mRNA | BT132752.1 | - | 11 - 12 | 6531 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
