Assay Details
| Gene Symbol: ck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299020.1 | NP_001285949.1 | 7 - 8 | 4965 | 86 | |
| NM_001299021.1 | NP_001285950.1 | 7 - 8 | 5128 | 86 | ||
| NM_078847.4 | NP_523571.1 | 7 - 8 | 4960 | 86 | ||
| NM_165099.3 | NP_723895.1 | 7 - 8 | 4965 | 86 | ||
| GenBank mRNA | BT132752.1 | - | 7 - 8 | 4912 | 86 | |
More Information
Set Membership: |
|

ck
Genomic Map