Assay Details
| Gene Symbol: CycE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259108.2 | NP_001246037.1 | 2 - 3 | 497 | 59 | |
| NM_057611.5 | NP_476959.1 | 3 - 4 | 787 | 59 | ||
| NM_165123.3 | NP_723924.1 | 3 - 4 | 625 | 59 | ||
| NM_165124.3 | NP_723925.1 | 4 - 5 | 877 | 59 | ||
| NM_165125.3 | NP_723926.1 | 3 - 4 | 634 | 59 | ||
| GenBank mRNA | AY069507.2 | - | 3 - 4 | 627 | 59 | |
More Information
Set Membership: |
|

CycE
Genomic Map