Assay Details
Gene Symbol: BicC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057517.3 | NP_476865.1 | 4 - 5 | 1242 | 76 | |
NM_165144.2 | NP_723948.1 | 3 - 4 | 919 | 76 | ||
GenBank mRNA | BT023837.1 | - | 4 - 5 | 1159 | 76 |
Gene Symbol: CG45084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299044.2 | NP_001285973.1 | 3 - 4 | 919 | 76 | |
NM_001299045.1 | NP_001285974.1 | 4 - 5 | 1242 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
