Assay Details
Gene Symbol: BicC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057517.3 | NP_476865.1 | 6 - 7 | 1390 | 63 | |
NM_165144.2 | NP_723948.1 | 5 - 6 | 1067 | 63 | ||
NM_165145.1 | NP_723949.1 | 2 - 3 | 386 | 63 | ||
GenBank mRNA | BT023837.1 | - | 6 - 7 | 1307 | 63 |
Gene Symbol: CG45084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299044.2 | NP_001285973.1 | 5 - 6 | 1067 | 63 | |
NM_001299045.1 | NP_001285974.1 | 6 - 7 | 1390 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
