Assay Details
Gene Symbol: Ca-alpha1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273551.1 | NP_001260480.2 | 23 - 24 | 5773 | 74 | |
NM_001273552.1 | NP_001260481.1 | 22 - 23 | 4686 | 74 | ||
NM_001273553.1 | NP_001260482.1 | 22 - 23 | 4686 | 74 | ||
NM_001273554.1 | NP_001260483.1 | 22 - 23 | 4686 | 74 | ||
NM_001273555.1 | NP_001260484.1 | 22 - 23 | 4695 | 74 | ||
NM_001273556.1 | NP_001260485.1 | 23 - 24 | 4794 | 74 | ||
NM_080365.3 | NP_525104.3 | 23 - 24 | 5778 | 74 | ||
NM_134429.3 | NP_602305.2 | 23 - 24 | 5769 | 74 | ||
NM_165146.2 | NP_723952.2 | 24 - 25 | 5791 | 74 | ||
NM_165147.2 | NP_723953.2 | 23 - 24 | 5683 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
