Assay Details
Gene Symbol: Ca-alpha1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273551.1 | NP_001260480.2 | 28 - 29 | 6829 | 87 | |
NM_001273552.1 | NP_001260481.1 | 27 - 28 | 5742 | 87 | ||
NM_001273553.1 | NP_001260482.1 | 27 - 28 | 5742 | 87 | ||
NM_001273554.1 | NP_001260483.1 | 27 - 28 | 5742 | 87 | ||
NM_001273555.1 | NP_001260484.1 | 27 - 28 | 5751 | 87 | ||
NM_001273556.1 | NP_001260485.1 | 28 - 29 | 5850 | 87 | ||
NM_080365.3 | NP_525104.3 | 28 - 29 | 6834 | 87 | ||
NM_134429.3 | NP_602305.2 | 28 - 29 | 6825 | 87 | ||
NM_165146.2 | NP_723952.2 | 29 - 30 | 6847 | 87 | ||
NM_165147.2 | NP_723953.2 | 28 - 29 | 6739 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
