Assay Details
| Gene Symbol: dac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014486.3 | NP_001014486.1 | 3 - 4 | 1695 | 75 | |
| NM_001014487.3 | NP_001014487.1 | 4 - 5 | 1722 | 75 | ||
| NM_165159.3 | NP_723968.1 | 3 - 4 | 1701 | 75 | ||
| NM_165160.3 | NP_723969.1 | 3 - 4 | 1722 | 75 | ||
| NM_165161.3 | NP_723970.1 | 4 - 5 | 1728 | 75 | ||
| NM_165162.3 | NP_723971.1 | 4 - 5 | 1749 | 75 | ||
More Information
Set Membership: |
|

dac
Genomic Map