Assay Details
| Gene Symbol: dac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014486.3 | NP_001014486.1 | 6 - 7 | 2910 | 65 | |
| NM_001014487.3 | NP_001014487.1 | 7 - 8 | 2937 | 65 | ||
| NM_165159.3 | NP_723968.1 | 6 - 7 | 2916 | 65 | ||
| NM_165160.3 | NP_723969.1 | 6 - 7 | 2937 | 65 | ||
| NM_165161.3 | NP_723970.1 | 7 - 8 | 2943 | 65 | ||
| NM_165162.3 | NP_723971.1 | 7 - 8 | 2964 | 65 | ||
More Information
Set Membership: |
|

dac
Genomic Map