Assay Details
Gene Symbol: Tpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273571.1 | NP_001260500.1 | 3 - 4 | 1455 | 76 | |
NM_001273572.1 | NP_001260501.1 | 3 - 4 | 1810 | 76 | ||
NM_001273573.1 | NP_001260502.1 | 4 - 5 | 1951 | 76 | ||
NM_078860.3 | NP_523584.1 | 3 - 4 | 1810 | 76 | ||
NM_165164.2 | NP_723974.1 | 3 - 4 | 1681 | 76 | ||
GenBank mRNA | AF199022.1 | - | 3 - 4 | 1803 | 76 | |
BT100099.1 | - | 3 - 4 | 1684 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
