Assay Details
Gene Symbol: glu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165180.2 | NP_723996.1 | 3 - 4 | 2187 | 83 | |
GenBank mRNA | AF186472.1 | - | 3 - 4 | 2165 | 83 | |
BT031125.1 | - | 3 - 4 | 2152 | 83 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
