Assay Details
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169519.2 | NP_001162990.1 | 14 - 15 | 3766 | 71 | |
NM_001169520.3 | NP_001162991.1 | 14 - 15 | 3928 | 71 | ||
NM_001259119.2 | NP_001246048.1 | 14 - 15 | 3766 | 71 | ||
NM_001259121.2 | NP_001246050.1 | 13 - 14 | 3716 | 71 | ||
NM_001259122.1 | NP_001246051.1 | 14 - 15 | 3926 | 71 | ||
NM_078863.6 | NP_523587.4 | 14 - 15 | 3766 | 71 | ||
NM_165181.3 | NP_723999.1 | 14 - 15 | 3766 | 71 | ||
NM_165182.3 | NP_724000.1 | 14 - 15 | 3766 | 71 | ||
NM_165183.3 | NP_724001.1 | 14 - 15 | 3766 | 71 | ||
NM_165184.3 | NP_724002.2 | 14 - 15 | 3908 | 71 | ||
NM_165185.3 | NP_724003.1 | 14 - 15 | 3766 | 71 | ||
NM_165186.3 | NP_724004.1 | 14 - 15 | 3766 | 71 | ||
NM_165187.3 | NP_724005.1 | 14 - 15 | 3766 | 71 | ||
NM_165188.3 | NP_724006.1 | 14 - 15 | 3766 | 71 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
