Assay Details
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169520.3 | NP_001162991.1 | 2 - 3 | 483 | 116 | |
NM_001259119.2 | NP_001246048.1 | 2 - 3 | 321 | 116 | ||
NM_001259120.1 | NP_001246049.1 | 2 - 3 | 476 | 116 | ||
NM_001316398.1 | NP_001303327.1 | 2 - 3 | 476 | 116 | ||
NM_165181.3 | NP_723999.1 | 2 - 3 | 321 | 116 | ||
NM_165182.3 | NP_724000.1 | 2 - 3 | 321 | 116 | ||
NM_165183.3 | NP_724001.1 | 2 - 3 | 321 | 116 | ||
NM_165184.3 | NP_724002.2 | 2 - 3 | 463 | 116 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
