Assay Details
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169519.2 | NP_001162990.1 | 6 - 7 | 1010 | 107 | |
NM_001169520.3 | NP_001162991.1 | 6 - 7 | 1172 | 107 | ||
NM_001259122.1 | NP_001246051.1 | 6 - 7 | 1170 | 107 | ||
NM_078863.6 | NP_523587.4 | 6 - 7 | 1010 | 107 | ||
NM_165181.3 | NP_723999.1 | 6 - 7 | 1010 | 107 | ||
NM_165182.3 | NP_724000.1 | 6 - 7 | 1010 | 107 | ||
NM_165183.3 | NP_724001.1 | 6 - 7 | 1010 | 107 | ||
NM_165185.3 | NP_724003.1 | 6 - 7 | 1010 | 107 | ||
NM_165186.3 | NP_724004.1 | 6 - 7 | 1010 | 107 | ||
NM_165187.3 | NP_724005.1 | 6 - 7 | 1010 | 107 | ||
NM_165188.3 | NP_724006.1 | 6 - 7 | 1010 | 107 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
