Assay Details
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169519.2 | NP_001162990.1 | 3 - 4 | 468 | 79 | |
NM_001169521.2 | NP_001162992.1 | 3 - 4 | 468 | 79 | ||
NM_001259121.2 | NP_001246050.1 | 2 - 3 | 418 | 79 | ||
NM_001259122.1 | NP_001246051.1 | 3 - 4 | 628 | 79 | ||
NM_078863.6 | NP_523587.4 | 3 - 4 | 468 | 79 | ||
NM_165185.3 | NP_724003.1 | 3 - 4 | 468 | 79 | ||
NM_165186.3 | NP_724004.1 | 3 - 4 | 468 | 79 | ||
NM_165187.3 | NP_724005.1 | 3 - 4 | 468 | 79 | ||
NM_165188.3 | NP_724006.1 | 3 - 4 | 468 | 79 | ||
NM_165189.3 | NP_724007.1 | 3 - 4 | 468 | 79 | ||
NM_165190.4 | NP_724008.1 | 3 - 4 | 636 | 79 | ||
NM_165191.3 | NP_724009.1 | 3 - 4 | 468 | 79 | ||
NM_165192.4 | NP_724010.1 | 3 - 4 | 468 | 79 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
