Assay Details
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169521.2 | NP_001162992.1 | 7 - 8 | 1120 | 69 | |
NM_165189.3 | NP_724007.1 | 7 - 8 | 1120 | 69 | ||
NM_165190.4 | NP_724008.1 | 7 - 8 | 1288 | 69 | ||
NM_165191.3 | NP_724009.1 | 7 - 8 | 1120 | 69 | ||
NM_165192.4 | NP_724010.1 | 7 - 8 | 1120 | 69 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
