Assay Details
Gene Symbol: shakB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103568.3 | NP_001097038.2 | 7 - 8 | 2267 | 93 | |
NM_001144750.2 | NP_001138222.1 | 6 - 7 | 1008 | 93 | ||
NM_001258858.2 | NP_001245787.1 | 10 - 11 | 2119 | 93 | ||
NM_001258859.2 | NP_001245788.1 | 11 - 12 | 2983 | 93 | ||
NM_001298574.1 | NP_001285503.1 | 10 - 11 | 3224 | 93 | ||
NM_078701.3 | NP_523425.2 | 9 - 10 | 2415 | 93 | ||
NM_134566.3 | NP_608410.2 | 8 - 9 | 1519 | 93 | ||
NM_170660.2 | NP_728361.1 | 6 - 7 | 1767 | 93 | ||
GenBank mRNA | S78495.1 | - | 6 - 7 | 1767 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
