Assay Details
| Gene Symbol: Plc21C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272889.1 | NP_001259818.1 | 6 - 7 | 1440 | 73 | |
| NM_057503.5 | NP_476851.2 | 6 - 7 | 1391 | 73 | ||
| NM_057504.5 | NP_476852.1 | 6 - 7 | 1440 | 73 | ||
| NM_205882.2 | NP_995604.2 | 6 - 7 | 1391 | 73 | ||
| NM_205883.2 | NP_995605.1 | 6 - 7 | 1391 | 73 | ||
| NM_205884.3 | NP_995606.1 | 6 - 7 | 1440 | 73 | ||
| GenBank mRNA | AY051657.1 | - | 6 - 7 | 1403 | 73 | |
More Information
Set Membership: |
|

Plc21C
Genomic Map