Assay Details
Gene Symbol: Plc21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057503.5 | NP_476851.2 | 8 - 9 | 1771 | 70 | |
NM_057504.5 | NP_476852.1 | 8 - 9 | 1820 | 70 | ||
NM_205882.2 | NP_995604.2 | 8 - 9 | 1771 | 70 | ||
NM_205883.2 | NP_995605.1 | 8 - 9 | 1771 | 70 | ||
NM_205884.3 | NP_995606.1 | 8 - 9 | 1820 | 70 | ||
GenBank mRNA | AY051657.1 | - | 8 - 9 | 1783 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
