Assay Details
| Gene Symbol: AP-2alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057471.6 | NP_476819.2 | 3 - 4 | 806 | 77 | |
| NM_205885.2 | NP_995607.1 | 2 - 3 | 373 | 77 | ||
| GenBank mRNA | BT032839.1 | - | 3 - 4 | 787 | 77 | |
| BT125866.1 | - | 2 - 3 | 373 | 77 | ||
| Y11104.1 | - | 675 | 77 | |||
More Information
Set Membership: |
|

AP-2alpha
Genomic Map