Assay Details
| Gene Symbol: Arpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135152.4 | NP_608996.1 | 1 - 2 | 625 | 62 | |
| GenBank mRNA | AY113475.1 | - | 1 - 2 | 619 | 62 | |
| BT125913.1 | - | 1 - 2 | 650 | 62 | ||
More Information
Set Membership: |
|

Arpc4
Genomic Map