Assay Details
Gene Symbol: mmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135188.3 | NP_609032.1 | 1 - 2 | 275 | 72 | |
GenBank mRNA | AF145686.1 | - | 1 - 2 | 260 | 72 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
