Assay Details
Gene Symbol: COX5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298749.1 | NP_001285678.1 | 2 - 3 | 357 | 59 | |
NM_135202.2 | NP_609046.1 | 2 - 3 | 357 | 59 | ||
GenBank mRNA | AY070965.1 | - | 2 - 3 | 355 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
