Assay Details
Gene Symbol: Proc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298799.1 | NP_001285728.1 | 2 - 3 | 381 | 61 | |
NM_135314.3 | NP_609158.2 | 2 - 3 | 472 | 61 | ||
GenBank mRNA | AJ458447.1 | - | 2 - 3 | 496 | 61 | |
AY119161.1 | - | 2 - 3 | 496 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
