Assay Details
| Gene Symbol: Npc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201840.2 | NP_001188769.1 | 10 - 11 | 3084 | 85 | |
| NM_001201841.2 | NP_001188770.1 | 10 - 11 | 3046 | 85 | ||
| NM_001201842.2 | NP_001188771.1 | 10 - 11 | 3012 | 85 | ||
| NM_001298866.1 | NP_001285795.1 | 10 - 11 | 3176 | 85 | ||
| NM_001298867.1 | NP_001285796.1 | 10 - 11 | 2997 | 85 | ||
| NM_001298868.1 | NP_001285797.1 | 10 - 11 | 3990 | 85 | ||
| NM_135513.5 | NP_609357.2 | 10 - 11 | 3188 | 85 | ||
More Information
Set Membership: |
|

Npc1a
Genomic Map