Assay Details
| Gene Symbol: Npc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201840.2 | NP_001188769.1 | 7 - 8 | 1085 | 66 | |
| NM_001201841.2 | NP_001188770.1 | 7 - 8 | 1047 | 66 | ||
| NM_001201842.2 | NP_001188771.1 | 7 - 8 | 1013 | 66 | ||
| NM_135513.5 | NP_609357.2 | 7 - 8 | 1189 | 66 | ||
More Information
Set Membership: |
|

Npc1a
Genomic Map