Assay Details
| Gene Symbol: Npc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201840.2 | NP_001188769.1 | 9 - 10 | 1697 | 89 | |
| NM_001201841.2 | NP_001188770.1 | 9 - 10 | 1659 | 89 | ||
| NM_001201842.2 | NP_001188771.1 | 9 - 10 | 1625 | 89 | ||
| NM_001298866.1 | NP_001285795.1 | 9 - 10 | 1789 | 89 | ||
| NM_001298867.1 | NP_001285796.1 | 9 - 10 | 1610 | 89 | ||
| NM_001298868.1 | NP_001285797.1 | 9 - 10 | 2603 | 89 | ||
| NM_135513.5 | NP_609357.2 | 9 - 10 | 1801 | 89 | ||
More Information
Set Membership: |
|

Npc1a
Genomic Map