Assay Details
Gene Symbol: Mob3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135520.4 | NP_609364.1 | 2 - 3 | 376 | 96 | |
GenBank mRNA | AY071000.1 | - | 2 - 3 | 375 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
